Order Entry
United States
ContactUsLinkComponent
Anti-POLG2 Mouse Polyclonal Antibody
Anti-POLG2 Mouse Polyclonal Antibody
Catalog # 103340-422
Supplier:  Avantor
Anti-POLG2 Mouse Polyclonal Antibody
Catalog # 103340-422
Supplier:  Avantor
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Symbol:
    POLG2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    0.05 mg
  • Environmentally Preferable:
  • Gene ID:
    11232
  • Antigen Synonyms:
    accessory subunit|PolG-beta|PEOA4|POLB|p55|EC 2.7.7.7|polymerase (DNA directed)|mitochondrial|MtPolB|gamma 2|mitochondrial DNA polymerase|MTPOLBPOLG-BETA|Mitochondrial DNA polymerase accessory subunit|DNA polymerase gamma accessory 55 kDa subunit|HP55|mitochondrial DNA polymerase subunit gamma-2|POLGB|DNA polymerase subunit gamma-2
  • Storage Buffer:
    PBS (pH 7.4)
  • Storage Temperature:
    Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
  • Immunogen:
    POLG2 (AAH09194, 1 a.a. ~ 485 a.a) full-length human protein.MRSRVAVRACHKVCRCLLSGFGGRVDAGQPELLTERSSPKGGHVKSHAELEGNGEHPEAPGSGEGSEALLEICQRRHFLSGSKQQLSRDSLLSGCHPGFGPLGVELRKNLAAEWWTSVVVFREQVFPVDALHHKPGPLLPGDSAFRLVSAETLREILQDKELSKEQLVAFLENVLKTSGKLRENLLHGALEHYVNCLDLVNKRLPYGLAQIGVCFHPVFDTKQIRNGVKSIGEKTEASLVWFTPPRTSNQWLDFW
  • Purification:
    Protein A purified
  • Cat. No.:
    103340-422
  • Supplier no.:
    H00011232-B01P

Specifications

About this item

The POLG2 Antibody from Novus Biologicals is a mouse polyclonal antibody to POLG2. This antibody reacts with human. The POLG2 Antibody has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: POLG2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human