To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:PLCL2
- Clonality:Monoclonal
- Clone:2D10
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:23228
- Antigen Synonyms:FLJ13484|phospholipase C|Phospholipase C-L2|PLC-L(2)|Phospholipase C-epsilon-2|epsilon 2|phospholipase C-like 2|EC 3.1.4.11|KIAA1092PLCE2|PLC-L2|inactive phospholipase C-like protein 2|PLC-epsilon-2
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:PLCL2 (AAH36392, 121 a.a. ~ 211 a.a) partial recombinant protein with GST tag.RYLISYGKHTLDMLESSQDNMRTSWVSQMFSEIDVDNLGHITLCNAVQCIRNLNPGLKTSKIELKFKELHKSKDKAGTEVTKEEFIEVFH
- Purification:IgG purified
- Cat. No.:103341-122
- Supplier no.:H00023228-M01
Specifications
About this item
The PLCL2 Antibody (2D10) from Novus Biologicals is a mouse monoclonal antibody to PLCL2. This antibody reacts with human. The PLCL2 Antibody (2D10) has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: PLCL2
Clonality: Monoclonal
Clone: 2D10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human