To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Symbol:p23/PTGES3
- Clonality:Monoclonal
- Clone:3C6-2B9
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:10728
- Antigen Synonyms:23 kD|Cytosolic prostaglandin E2 synthase|Telomerase-binding protein p23|prostaglandin E synthase 3 (cytosolic)|Progesterone receptor complex p23|TEBPEC 5.3.99.3|cPGESp23|unactive progesterone receptor|Hsp90 co-chaperone|prostaglandin E synthase 3|P23cytosolic prostaglandin E synthase
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:PTGES3 (AAH03005, 1 a.a. ~ 161 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.MQPASAKWYDRRDYVFIEFCVEDSKDVNVNFEKSKLTFSCLGGSDNFKHLNEIDLFHCIDPNDSKHKRTDRSILCCLRKGESGQSWPRLTKERAKLNWLSVDFNNWKDWEDDSDEDMSNFDRFSEMMNNMGGDEDVDLPEVDGADDDSQDSDDEKMPDLE
- Purification:IgG purified
- Cat. No.:103339-496
- Supplier no.:H00010728-M02
The p23 / PTGES3 Antibody (3C6-2B9) from Novus Biologicals is a mouse monoclonal antibody to p23 / PTGES3. This antibody reacts with human. The p23 / PTGES3 Antibody (3C6-2B9) has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: p23/PTGES3
Clonality: Monoclonal
Clone: 3C6-2B9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human