Anti-NSD3 Rabbit Polyclonal Antibody
100 μl
Catalog # 103279-908
Marketsource
Supplier: Novus Biologicals
Supplier #: NBP1-89852
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
The NSD3 Antibody from Novus Biologicals is a rabbit polyclonal antibody to NSD3. This antibody reacts with human. The NSD3 Antibody has been validated for the following applications: Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: NSD3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Size:100 μl
- Clonality:Polyclonal
- Purification:Immunogen affinity purified
- Storage Buffer:PBS (pH 7.2) and 40% Glycerol
- ImmunoFluorescence:Yes
- Isotype:IgG
- Host:Rabbit
- Cat. No.:103279-908
- Reactivity:Human
- Antibody Type:Primary
- Antigen Symbol:NSD3
- Gene ID:54904
- Immunogen:This antibody was developed against Recombinant Protein corresponding to amino acids:MMRCLRCPVAYHSGDACIAAGSMLVSSYILICSNHSKRSSNSSAVNVGFCFVCARGLIVQDHSDPMFSSYAYKSHYLLNESNRAELMKLPMIPSSSASKKKCEKGGRLLCCESCPASFHPECLSIEMPEGCWNC
- ImmunoChemistry:Yes
- Conjugation:Unconjugated
- Antigen Synonyms:Wolf-Hirschhorn syndrome candidate 1-like protein 1|histone-lysine N-methyltransferase NSD3|MGC142029|WHSC1-like protein 1|MGC126766|Nuclear SET domain-containing protein 3|pp14328|Protein whistle|NSD3DKFZp667H044|FLJ20353|EC 2.1.1.43|Wolf-Hirschhorn syndrome candidate 1-like 1|WHSC1-like 1 isoform 9 with methyltransferase activity to lysine
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



