To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:Nogo
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:0.2 ml
- Environmentally Preferable:
- Gene ID:57142
- Antigen Synonyms:reticulon-5|Nbla00271|Nogo-B|RTN-x|Neuroendocrine-specific protein|NSP-CL|NOGO-A|NI220/250|My043 protein|RTN4-A|RTN4-C|Nbla10545|RTN4-B2|Nogo protein|neurite growth inhibitor 220|Nogo-C|NSP|NOGOC|reticulon 4|Human NogoA|Neurite outgrowth inhibitor|KIAA0886|NOGORTN4-B1|reticulon 5|foocen|Neuroendocrine-specific protein C homolog|reticulon-4|ASY
- Storage Buffer:PBS
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Concentration:1.0 mg/ml
- Immunogen:A synthetic peptide to an internal portion of human Nogo A (between amino acids 1-50) [UniProt Q9NQC3].
- Tested Applications:Simple Western
- Purification:Protein G purified
- Cat. No.:103361-088
Specifications
About this item
The Nogo Antibody from Novus Biologicals is a rabbit polyclonal antibody to Nogo. This antibody reacts with human, mouse, rat. The Nogo Antibody has been validated for the following applications: Western Blot, Simple Western, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: Nogo
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat