To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Symbol:NM23-H2/NME2
- Clonality:Monoclonal
- Clone:1A9
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG3 kappa
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:4831
- Antigen Synonyms:NDPK-B|non-metastatic cells 2|protein (NM23B) expressed in|Histidine protein kinase NDKB|nm23-H2|NDK B|protein (NM23) expressed in|NDKB|NDPKB|EC 2.7.13.3|MGC2212|nucleoside diphosphate kinase B
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:NME2 (NP_002503, 51 a.a. - 152 a.a.) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. HYIDLKDRPFFPGLVKYMNSGPVVAMVWEGLNVVKTGRVMLGETNPADSKPGTIRGDFCIQVGRNIIHGSDSVKSAEKEISLWFKPEELVDYKSCAHDWVYE
- Purification:IgG purified
- Cat. No.:103331-780
- Supplier no.:H00004831-M09
The NM23-H2 / NME2 Antibody (1A9) from Novus Biologicals is a mouse monoclonal antibody to NM23-H2 / NME2. This antibody reacts with human, mouse, rat. The NM23-H2 / NME2 Antibody (1A9) has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence.
Type: Primary
Antigen: NM23-H2/NME2
Clonality: Monoclonal
Clone: 1A9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3 Kappa
Reactivity: Human, Mouse, Rat