To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Symbol:NM23-H1
- Clonality:Monoclonal
- Clone:2H1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:4830
- Antigen Synonyms:EC 2.7.4.6|nm23-H1|NDPKA|Tumor metastatic process-associated protein|NDKA|non-metastatic cells 1|NBS|protein (NM23A) expressed in|NDP kinase A|Granzyme A-activated DNase|GAAD|nucleoside diphosphate kinase A|NDK A|NDPK-A|NM23AWD|Metastasis inhibition factor nm23|NB
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:NME1 (AAH00293, 1 a.a. - 152 a.a.) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MANCERTFIAIKPDGVQRGLVGEIIKRFEQKGFRLVGLKFMQASEDLLKEHYVDLKDRPFFAGLVKYMHSGPVVAMVWEGLNVVKTGRVMLGETNPADSKPGTIRGDFCIQVGRNIIHGSDSVESAEKEIGLWFHPEELVDYTSCAQNWIYE
- Tested Applications:, RNA Inhibition
- Purification:IgG purified
- Cat. No.:103331-770
- Supplier no.:H00004830-M01
The NM23-H1 Antibody (2H1) from Novus Biologicals is a mouse monoclonal antibody to NM23-H1. This antibody reacts with human. The NM23-H1 Antibody (2H1) has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence, RNA Inhibition.
Type: Primary
Antigen: NM23-H1
Clonality: Monoclonal
Clone: 2H1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human