To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Symbol:NEDP1/SENP8
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 ug
- Environmentally Preferable:
- Gene ID:123228
- Antigen Synonyms:2 (NEDD8 specific)|Sentrin/SUMO-specific protease SENP8|HsT17512|EC 3.4.22.-|deneddylase 1|cysteine|cysteine 2|NEDP1NEDD8 specific-protease cysteine 2|sentrin-specific protease 8|Deneddylase-1|protease|DEN1deneddylase-1|SUMO/sentrin specific protease family member 8|SUMO/sentrin specific peptidase family member 8|SUMO sentrin specific protease family member 8|NEDD8-specific protease 1|PRSC2|Protease
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:SENP8 (NP_660205.2, 1 a.a. ~ 212 a.a) full-length human protein.MDPVVLSYMDSLLRQSDVSLLDPPSWLNDHIIGFAFEYFANSQFHDCSDHVSFISPEVTQFIKCTSNPAEIAMFLEPLDLPNKRVVFLAINDNSNQAAGGTHWSLLVYLQDKNSFFHYDSHSRSNSVHAKQVAEKLEAFLGRKGDKLAFVEEKAPAQQNSYDCGMYVICNTEALCQNFFRQQTESLLQLLTPAYITKKRGEWKDLIATLAKK
- Purification:Protein G purified
- Cat. No.:103346-544
- Supplier no.:H00123228-B01P
The NEDP1 / SENP8 Antibody from Novus Biologicals is a mouse polyclonal antibody to NEDP1 / SENP8. This antibody reacts with human. The NEDP1 / SENP8 Antibody has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence.
Type: Primary
Antigen: NEDP1/SENP8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human