ContactUsLinkComponent
CLINITEST® hCG Pregnancy Test, Siemens Healthineers
CLINITEST® hCG Pregnancy Test, Siemens Healthineers
Catalog # 89140-208
CAS Number:  
undefined
CLINITEST® hCG Pregnancy Test, Siemens Healthineers
Catalog # 89140-208
Supplier Number:  10310618
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Clinitest® hCG Pregnancy Test, CLIA Waived
  • Environmentally Preferable:
  • Cat. No.:
    89140-208
  • Parameters tested:
    hCG
  • Supplier no.:
    1760
Specifications
About this item
CLINITEST® hCG Pregnancy Test from Siemens Healthineers gives you fast, easy instrument-read-and-reported test results for early detection of pregnancy.
  • Ideal for busy point-of-care testing sites
  • Reduced user involvement and time
  • Enhanced uniform test method
  • Decreased user uncertainty
  • Less opportunity for error
  • More objective results
  • Increased time for patient care
  • Consistent, reliable results every time
  • CLIA-waived
Qualitative, instrument-read test for early detection of pregnancy is for use on the CLINITEK Status family of analyzers. Provides a flag for results in the "indeterminate" range where confirmatory testing is important for interpreting significance of low level hCG in qualitative tests. Quick, reliable instrument-read results eliminate interpretation and provide accurate results for your patient. The CLINITEK Status+ Analyzer precisely times the reading of results, eliminating hands-on waiting time. Printed results eliminate transcription errors and save time. Sensitivity equal to or greater than 25 mIU/mL with clinical accuracy of >99%.