To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:MST4
- Clonality:Monoclonal
- Clone:2G6
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2b kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:51765
- Antigen Synonyms:MASK|Serine/threonine-protein kinase MASK|mammalian sterile 20-like 4|serine/threonine-protein kinase MST4|Mst3 and SOK1-related kinase|MST-4|serine/threonine protein kinase MST4|serine/threonine protein kinase MASK|STE20-like kinase MST4|EC 2.7.11.1|Mammalian STE20-like protein kinase 4|EC 2.7.11
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:RP6-213H19.1 (AAH17213, 1 a.a. ~ 138 a.a) full length recombinant protein with GST tag.MAHSPVAVQVPGMQNNIADPEELFTKLERIGKGSFGEVFKGI DNRTQQVVAIKIIDLEEAEDEIEDIQQEITVLSQCDSSYVTK YYGSYLKGSKLWIIMEYLGGGSALDLLRALPPYERSLIQRKY RMGQSKILCKP
- Purification:IgG purified
- Cat. No.:103344-492
- Supplier no.:H00051765-M01
Specifications
About this item
The MST4 Antibody (2G6) from Novus Biologicals is a mouse monoclonal antibody to MST4. This antibody reacts with human. The MST4 Antibody (2G6) has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence.
Type: Primary
Antigen: MST4
Clonality: Monoclonal
Clone: 2G6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human