To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:MCM9
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:254394
- Antigen Synonyms:MGC35304|DNA replication licensing factor MCM9|FLJ56845|FLJ20170|dJ329L24.3|minichromosome maintenance complex component 9|FLJ13942|dJ329L24.1|Mini-chromosome maintenance deficient domain-containing protein 1|hMCM9|MCMDC1|minichromosome maintenance deficient domain containing 1|Mini-chromosome maintenance deficient 9|chromosome 6 open reading frame 61|C6orf61
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:MCM9 (NP_694987.1, 1 a.a. ~ 391 a.a) full-length human protein.
- Purification:Protein G purified
- Cat. No.:103347-962
- Supplier no.:H00254394-D01P
Specifications
About this item
The MCM9 Antibody from Novus Biologicals is a rabbit polyclonal antibody to MCM9. This antibody reacts with human, rat. The MCM9 Antibody has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: MCM9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat