To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:MBD1
- Clonality:Monoclonal
- Clone:100B272.1
- Conjugation:HRP (Horseradish Peroxidase)
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:4152
- Antigen Synonyms:the regulator of fibroblast growth factor 2 (FGF-2) transcription|PCM1CXXC3RFT|Methyl-CpG-binding protein MBD1|methyl-CpG-binding domain protein 1|CXXC-type zinc finger protein 3|methyl-CpG binding domain protein 1 isoform PCM1|Protein containing methyl-CpG-binding domain 1|methyl-CpG binding domain protein 1
- Storage Buffer:PBS
- Storage Temperature:Store at 4C in the dark.
- Immunogen:This antibody was generated by immunizing mice with a synthetic peptide corresponding to amino acids 391-405 (SESEDGAGSPPPYRR) of human MBD1; GenBank no ref NP_056671.2
- Purification:Protein G purified
- Cat. No.:103359-938
Specifications
About this item
The MBD1 Antibody (100B272.1) [HRP] from Novus Biologicals is a mouse monoclonal antibody to MBD1. This antibody reacts with human, mouse. The MBD1 Antibody (100B272.1) [HRP] has been validated for the following applications: Western Blot.
Type: Primary
Antigen: MBD1
Clonality: Monoclonal
Clone: 100B272.1
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse