To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:Lgr6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:0.05 ml
- Environmentally Preferable:
- Epitope:N-terminal extracellular domain of human
- Gene ID:59352
- Antigen Synonyms:VTS20631|gonadotropin receptor|leucine-rich repeat-containing G-protein coupled receptor 6|GPCR|leucine-rich repeat-containing G protein-coupled receptor 6|leucine-rich repeat containing G protein-coupled receptor 6|FLJ14471
- Storage Buffer:PBS
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Concentration:1.0 mg/ml
- Immunogen:Synthetic 20 amino acid peptide from N-terminal extracellular domain of human LGR6.
- Purification:Immunogen affinity purified
- Cat. No.:103264-056
- Supplier no.:NLS442
Specifications
About this item
The Lgr6 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Lgr6. This antibody reacts with human. The Lgr6 Antibody has been validated for the following applications: Immunohistochemistry-Paraffin.
Type: Primary
Antigen: Lgr6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal extracellular domain of human
Host: Rabbit
Isotype: IgG
Reactivity: Human