To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:LBX1
- Clonality:Monoclonal
- Clone:2F8
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:10660
- Antigen Synonyms:ladybird homeobox homolog 1|HPX6|LBX1Hlady bird-like homeobox|transcription factor similar to D. melanogaster homeodomain protein lady birdlate|Ladybird homeobox protein homolog 1|ladybird homeobox homolog 1 (Drosophila)|HPX-6|homeobox|transcription factor LBX1|ladybird homeobox 1
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:LBX1 (NP_006553, 133 a.a. ~ 219 a.a) full length recombinant protein with GST tag.
- Purification:IgG purified
- Cat. No.:103339-380
- Supplier no.:H00010660-M03
Specifications
About this item
The LBX1 Antibody (2F8) from Novus Biologicals is a mouse monoclonal antibody to LBX1. This antibody reacts with human. The LBX1 Antibody (2F8) has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: LBX1
Clonality: Monoclonal
Clone: 2F8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human