To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Kallikrein 4
- Antigen Symbol:KLK4
- Clonality:Monoclonal
- Clone:2A4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:9622
- Antigen Synonyms:EC 3.4.21.-|kallikrein 4 (prostase|enamel matrix|EC 3.4.21|Enamel matrix serine proteinase 1|EMSP|ARM1|PRSS17enamel matrix serine protease 1|AI2A1|EMSP1MGC116827|Prostase|prostate)|kallikrein-4|Serine protease 17|Kallikrein-like protein 1|kallikrein-related peptidase 4|PSTSandrogen-regulated message 1|KLK-L1MGC116828
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:KLK4 (NP_004908, 159 a.a. ~ 254 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26. GRMPTVLQCVNVSVVSEEVCSKLYDPLYHPSMFCAGGGQDQKDSCNGDSGGPLICNGYLQGLVSFGKAPCGQVGVPGVYTNLCKFTEWIEKTVQAS
- Purification:IgG purified
- Cat. No.:103337-370
- Supplier no.:H00009622-M09
Specifications
About this item
The Kallikrein 4 / Prostase / EMSP1 Antibody (2A4) from Novus Biologicals is a mouse monoclonal antibody to Kallikrein 4 / Prostase / EMSP1. This antibody reacts with human. The Kallikrein 4 / Prostase / EMSP1 Antibody (2A4) has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: KLK4
Clonality: Monoclonal
Clone: 2A4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human