To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Glutamate Receptor KA2
- Antigen Symbol:KA2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human,Bovine,Rat
- Size:100 μg
- Environmentally Preferable:
- Epitope:C-Terminus
- Gene ID:2901
- Antigen Synonyms:Excitatory amino acid receptor 2|glutamate receptor KA2|EAA2|glutamate receptor|ionotropic|kainate 5|Glutamate receptor KA-2|KA2GRIK2|ionotropic kainate 5
- Storage Buffer:0.5 mg/ml Tris (pH 7.3) and 0.5% BSA
- Storage Temperature:Store at -20C. Avoid freeze-thaw cycles.
- Concentration:0.5 mg/ml
- Immunogen:Peptide with sequence C-GPAGPRELAEHE corresponding to C-Terminus according to NP_002079.3.
- Tested Applications:Peptide
- Purification:Immunogen affinity purified
- Cat. No.:103408-726
Specifications
About this item
The KA2 / GRIK5 / Glutamate Receptor KA2 Antibody from Novus Biologicals is a goat polyclonal antibody to KA2 / GRIK5 / Glutamate Receptor KA2. This antibody reacts with human, rat, bovine. The KA2 / GRIK5 / Glutamate Receptor KA2 Antibody has been validated for the following applications: Peptide ELISA.
Type: Primary
Antigen: KA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminus
Host: Goat
Isotype: IgG
Reactivity: Human, Rat, Bovine