To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:IL-27 R alpha
- Antigen Symbol:IL-27RA
- Clonality:Monoclonal
- Clone:8G9
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:9466
- Antigen Synonyms:zcytor1|alpha|IL27R|Cytokine receptor-like 1|interleukin 27 receptor|IL-27R-alpha|interleukin-27 receptor subunit alpha|class I cytokine receptor|Type I T-cell cytokine receptor|WSX1IL-27R subunit alpha|IL-27RA|T-cell cytokine receptor type 1|TCCRIL-27 receptor subunit alpha|WSX-1|CRL1IL-27R
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:IL27RA (NP_004834, 33 a.a. ~ 133 a.a) partial recombinant protein with GST tag.QGSAGPLQCYGVGPLGDLNCSWEPLGDLGAPSELHLQSQKYRSNKTQTVAVAAGRSWVAIPREQLTMSDKLLVWGTKAGQPLWPPVFVNLETQMKPNAPR
- Purification:IgG purified
- Cat. No.:103337-018
- Supplier no.:H00009466-M01
Specifications
About this item
The IL-27 R alpha / WSX-1 / TCCR Antibody (8G9) from Novus Biologicals is a mouse monoclonal antibody to IL-27 R alpha / WSX-1 / TCCR. This antibody reacts with human. The IL-27 R alpha / WSX-1 / TCCR Antibody (8G9) has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: IL-27RA
Clonality: Monoclonal
Clone: 8G9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human