To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:IL-13 R alpha 1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:3597
- Antigen Synonyms:IL-13RA1|alpha 1|IL-13R subunit alpha-1|Cancer/testis antigen 19|CD213a1 antigen|alpha 1)|bB128O4.2.1 (interleukin 13 receptor|IL-13R-alpha-1|interleukin 13 receptor|IL13 receptor alpha-1 chain|CD213A1|interleukin-13 receptor subunit alpha-1|IL13R|IL13RA|CT19|IL-13 receptor subunit alpha-1|NR4|IL-13Ra
- Storage Buffer:PBS (pH 7.0), 1.0% BSA and 20% Glycerol
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Concentration:0.18 mg/ml
- Immunogen:Recombinant protein encompassing a sequence within the center region of human IL13 Receptor alpha 1. The exact sequence is proprietary.
- Purification:Immunogen affinity purified
- Cat. No.:102708-836
- Supplier no.:NBP2-16942
Specifications
About this item
The IL-13 R alpha 1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to IL-13 R alpha 1. This antibody reacts with human. The IL-13 R alpha 1 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: IL-13 R alpha 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human