To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:IL-12 R beta 2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:3595
- Antigen Synonyms:IL-12RB2|interleukin-12 receptor subunit beta-2|IL-12R subunit beta-2|IL-12 receptor beta 2|interleukin-12 receptor beta-2 chain|IL-12R-beta-2|beta 2|interleukin 12 receptor|IL-12 receptor subunit beta-2
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:IL12RB2 (NP_001550.1, 1 a.a. - 862 a.a.) full-length human protein. MAHTFRGCSLAFMFIITWLLIKAKIDACKRGDVTVKPSHVILLGSTVNITCSLKPRQGCFHYSRRNKLILYKFDRRINFHHGHSLNSQVTGLPLGTTLFVCKLACINSDEIQICGAEIFVGVAPEQPQNLSCIQKGEQGTVACTWERGRDTHLYTEYTLQLSGPKNLTWQKQCKDIYCDYLDFGINLTPESPESNFTAKVTAVNSLGSSSSLPSTFTFLDIVRPLPPWDIRIKFQKASVSRCTLYWRDEGLVLLNRLRYRPSNSRLWNMVNVTKAKGRHDLLDLKPFTEYEFQISSKLHLYKGSWSDWSESLRAQTPEEEPTGMLDVWYMKRHIDYSRQQISLFWKNLSVSEARGKILHYQVTLQELTGGKAMTQNITGHTSWTTVIPRTGNWAVAVSAANSKGSSLPTRINIMNLCEAGLLAPRQVSANSEGMDNILVTWQPPRKDPSAVQEYVVEWRELHPGGDTQVPLNWLRSRPYNVSALISENIKSYICYEIRVYALSGDQGGCSSILGNSKHKAPLSGPHINAITEEKGSILISWNSIPVQEQMGCLLHYRIYWKERDSNSQPQLCEIPYRVSQNSHPINSLQPRVTYVLWMTALTAAGESSHGNEREFCLQGKANWMAFVAPSICIAIIMVGIFSTHYFQQKVFVLLAALRPQWCSREIPDPANSTCAKKYPIAEEKTQLPLDRLLIDWPTPEDPEPLVISEVLHQVTPVFRHPPCSNWPQREKGIQGHQASEKDMMHSASSPPPPRALQAESRQLVDLYKVLESRGSDPKPENPACPWTVLPAGDLPTHDGYLPSNIDDLPSHEAPLADSLEELEPQHISLSVFPSSSLHPLTFSCGDKLTLDQLKMRCDSLML
- Purification:Protein A purified
- Cat. No.:103329-532
- Supplier no.:H00003595-D01P
Specifications
About this item
The IL-12 R beta 2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to IL-12 R beta 2. This antibody reacts with human. The IL-12 R beta 2 Antibody has been validated for the following applications: Western Blot.
Type: Primary
Antigen: IL-12 R beta 2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human