To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:IL-1 RAcP/IL-1 R3
- Clonality:Monoclonal
- Clone:3D8
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Size:100 μg
- Environmentally Preferable:
- Gene ID:3556
- Antigen Synonyms:IL-1 receptor accessory protein|C3orf13IL-1RAcPIL1R3Interleukin-1 receptor 3|FLJ37788|IL-1R3|IL-1R-3|interleukin-1 receptor accessory protein beta|interleukin 1 receptor accessory protein|interleukin-1 receptor accessory protein
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:IL1RAP (AAH53621.1, 1 a.a. - 356 a.a.) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MTLLWCVVSLYFYGILQSDASERCDDWGLDTMRQIQVFEDEPARIKCPLFEHFLKFNYSTAHSAGLTLIWYWTRQDRDLEEPINFRLPENRISKEKDVLWFRPTLLNDTGNYTCMLRNTTYCSKVAFPLEVVQKDSCFNSPMKLPVHKLYIEYGIQRITCPNVDGYFPSSVKPTITWYMGCYKIQNFNNVIPEGMNLSFLIALISNNGNYTCVVTYPENGRTFHLTRTPTVKVVGSPKNAVPPVIHSPNDHVVYEKEPGEELLIPCTVYFSFLMDSRNEVWWTIDGKKPDDITIDVTINESISHSRTEDETRTQILSIKKVTSEDLKRSYVCHARSAKGEVAKAAKVKQKGNRCGQ
- Purification:IgG purified
- Cat. No.:103328-756
- Supplier no.:H00003556-M03
Specifications
About this item
The IL-1 RAcP / IL-1 R3 Antibody (3D8) from Novus Biologicals is a mouse monoclonal antibody to IL-1 RAcP / IL-1 R3. This antibody reacts with human. The IL-1 RAcP / IL-1 R3 Antibody (3D8) has been validated for the following applications: ELISA.
Type: Primary
Antigen: IL-1 RAcP/IL-1 R3
Clonality: Monoclonal
Clone: 3D8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human