To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Interferon C
- Antigen Symbol:IFNAC
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:3446
- Antigen Synonyms:interferon|alpha 10|interferon alpha-10|MGC119879|IFN-alpha-10|Interferon alpha-C|IFN-alphaC|Interferon alpha-6L|LeIF C|MGC119878
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:IFNA10 (NP_002162.1, 1 a.a. - 189 a.a.) full-length human protein. MALSFSLLMAVLVLSYKSICSLGCDLPQTHSLGNRRALILLGQMGRISPFSCLKDRHDFRIPQEEFDGNQFQKAQAISVLHEMIQQTFNLFSTEDSSAAWEQSLLEKFSTELYQQLNDLEACVIQEVGVEETPLMNEDSILAVRKYFQRITLYLIERKYSPCAWEVVRAEIMRSLSFSTNLQKRLRRKD
- Purification:Protein A purified
- Cat. No.:103328-934
- Supplier no.:H00003446-D01P
Specifications
About this item
The IFN-alpha C / IFN-alpha 10 Antibody from Novus Biologicals is a rabbit polyclonal antibody to IFN-alpha C / IFN-alpha 10. This antibody reacts with human. The IFN-alpha C / IFN-alpha 10 Antibody has been validated for the following applications: Western Blot.
Type: Primary
Antigen: IFNAC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human