Anti-HYPE Mouse Polyclonal Antibody
0.05 mg
Catalog # 103340-268
Marketsource
Jump to:
Discontinued
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
The HYPE Antibody from Novus Biologicals is a mouse polyclonal antibody to HYPE. This antibody reacts with human. The HYPE Antibody has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: HYPE
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Specifications
- Size:0.05 mg
- Clonality:Polyclonal
- Purification:Protein A purified
- Storage Buffer:PBS (pH 7.4)
- Isotype:IgG
- Host:Mouse
- Cat. No.:103340-268
- Reactivity:Human
- Western Blot:Yes
- Antibody Type:Primary
- Antigen Symbol:HYPE
- Gene ID:11153
- Immunogen:FICD (NP_009007.2, 1 a.a. ~ 458 a.a) full-length human protein.MMLIPMASVMAVTEPKWVSVWSRFLWVTLLSMVLGSLLALLLPLGAVEEQCLAVLKGLYLLRSKPDRAQHAATKCTSPSTELSITSRGATLLVAKTKASPAGKLEARAALNQALEMKRQGKREKAQKLFMHALKMDPDFVDALTEFGIFSEEDKDIIQADYLYTRALTISPYHEKALVNRDRTLPLVEEIDQRYFSIIDSKVKKVMSIPKGNSALRRVMEETYYHHIYHTVAIEGNTLTLSEIRHILETRYAVPG
- Conjugation:Unconjugated
- Antigen Synonyms:Huntingtin-interacting protein 13|HIP13UNQ3041|AMPylator FICD|Huntingtin-interacting protein E|MGC5623|fic S-phase protein cell division homolog|FIC domain containing|huntingtin interactor protein E|EC 2.7.7.n1|huntingtin interacting protein 13|Huntingtin interacting protein E|Huntingtin yeast partner E|FIC domain-containing protein|HIP-13|HYPEadenosine monophosphate-protein transferase FICD
- ELISA:Yes
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



