To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:HLA DPA
- Clonality:Monoclonal
- Clone:1E3.
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:3113
- Antigen Synonyms:HLA-SB alpha chain|MHC class II HLA-DPA1 antigen|MHC class II DPA1|Primed lymphocyte test-1|MHC class II DP3-alpha|DP alpha chain|major histocompatibility complex|class II|DP alpha 1|MHC class II antigen|DP alpha 1 chain|HLADP|HLA class II histocompatibility antigen
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:HLA-DPA1 (AAH09956, 1 a.a. - 260 a.a.) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MRPEDRMFHIRAVILRALSLAFLLSLRGAGAIKADHVSTYAAFVQTHRPTGEFMFEFDEDEQFYVDLDKKETVWHLEEFGRAFSFEAQGGLANIAILNNNLNTLIQRSNHTQAANDPPEVTVFPKEPVELGQPNTLICHIDRFFPPVLNVTWLCNGEPVTEGVAESLFLPRTDYSFHKFHYLTFVPSAEDVYDCRVEHWGLDQPLLKHWEAQEPIQMPETTETVLCALGLVLGLVGIIVGTVLIIKSLRSGHDPRAQGPL
- Purification:IgG purified
- Cat. No.:103328-120
- Supplier no.:H00003113-M03
Specifications
About this item
The HLA DPA Antibody (1E3.) from Novus Biologicals is a mouse monoclonal antibody to HLA DPA. This antibody reacts with human. The HLA DPA Antibody (1E3.) has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: HLA DPA
Clonality: Monoclonal
Clone: 1E3.
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human