Order Entry
United States
ContactUsLinkComponent
Anti-HIF-2 alpha/EPAS1 Rabbit Polyclonal Antibody (Alexa Fluor® 700)
Anti-HIF-2 alpha/EPAS1 Rabbit Polyclonal Antibody (Alexa Fluor® 700)
Catalog # 103367-972
Supplier:  Novus Biologicals
Anti-HIF-2 alpha/EPAS1 Rabbit Polyclonal Antibody (Alexa Fluor® 700)
Catalog # 103367-972
Supplier:  Novus Biologicals
Supplier Number:  NB100-122AF700
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Symbol:
    HIF-2 alpha/EPAS1
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 700
  • ELISA:
    Yes
  • Flow Cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Primates,
    Fish,
    Hamster,
    Sheep,
    Human,
    Rat,
    Mouse
  • Western Blot:
    Yes
  • Size:
    100 μl
  • Environmentally Preferable:
  • Gene ID:
    2034
  • Antigen Synonyms:
    Basic-helix-loop-helix-PAS protein MOP2|HIF2A|HIF2AHIF-2-alpha|Class E basic helix-loop-helix protein 73|hif2a angiogenesis|HIF-1alpha-like factor|bHLHe73ECYT4|endothelial PAS domain-containing protein 1|HLFHIF-1-alpha-like factor|EPAS-1|HIF-2 alpha|hypoxia-inducible factor 2 alpha|Member of PAS protein 2|MOP2HIF2-alpha|endothelial PAS domain protein 1|BHLHE73|PASD2PAS domain-containing protein 2|Hypoxia-inducible factor 2-alpha
  • Storage Buffer:
    50mM Sodium Borate
  • Storage Temperature:
    Store at 4C in the dark.
  • Immunogen:
    A peptide derived from the C-terminus of mouse/human HIF-2 alpha protein.
  • Purification:
    Immunogen affinity purified
  • Cat. No.:
    103367-972

Specifications

About this item

The HIF-2 alpha / EPAS1 Antibody [Alexa Fluor« 700] from Novus Biologicals is a rabbit polyclonal antibody to HIF-2 alpha / EPAS1. This antibody reacts with human, mouse, rat, fish, hamster, primate, sheep. The HIF-2 alpha / EPAS1 Antibody [Alexa Fluor« 700] has been validated for the following applications: Western Blot, Flow Cytometry, ELISA, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Type: Primary
Antigen: HIF-2 alpha/EPAS1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 700
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Fish, Hamster, Primate, Sheep