Order Entry
United States
ContactUsLinkComponent
Anti-HGK/MAP4K4 Rabbit Polyclonal Antibody
Anti-HGK/MAP4K4 Rabbit Polyclonal Antibody
Catalog # 103266-796
Supplier:  Novus Biologicals
Anti-HGK/MAP4K4 Rabbit Polyclonal Antibody
Catalog # 103266-796
Supplier:  Novus Biologicals
Supplier Number:  NBP1-84663
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Symbol:
    HGK/MAP4K4
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Size:
    100 μl
  • Environmentally Preferable:
  • Gene ID:
    9448
  • Antigen Synonyms:
    HPK/GCK-like kinase HGK|MEKKK 4|FLH21957|FLJ10410|EC 2.7.11.1|Nck-interacting kinase|mitogen-activated protein kinase kinase kinase kinase 4|NIKFLJ90111|KIAA0687|EC 2.7.11|MAPK/ERK kinase kinase kinase 4|MEK kinase kinase 4|hepatocyte progenitor kinase-like/germinal center kinase-like kinase|HGKFLJ20373
  • Storage Buffer:
    PBS (pH 7.2) and 40% Glycerol
  • Storage Temperature:
    Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
  • Immunogen:
    This antibody was developed against Recombinant Protein corresponding to amino acids:AEPQVQWSHLASLKNNVSPVSRSHSFSDPSPKFAHHHLRSQDPCPPSRSEVLSQSSDSKSEAPDPTQKAWSRSDSDEVPPRVPVRTTSRSPVLSRRDSPLQGSGQQNSQAGQRNSTSSI
  • Purification:
    Immunogen affinity purified
  • Cat. No.:
    103266-796
  • Supplier no.:
    NBP1-84663

Specifications

About this item

The HGK / MAP4K4 Antibody from Novus Biologicals is a rabbit polyclonal antibody to HGK / MAP4K4. This antibody reacts with human. The HGK / MAP4K4 Antibody has been validated for the following applications: Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: HGK/MAP4K4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human