To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:gtf3c3
- Clonality:Monoclonal
- Clone:3D9
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:9330
- Antigen Synonyms:polypeptide 3|Transcription factor IIIC 102 kDa subunit|Transcription factor IIIC subunit gamma|TFiiiC2-102|general transcription factor IIIC|polypeptide 3 (102kD)|TF3C-gamma|general transcription factor 3C polypeptide 3|transcription factor IIIC|102kDa|TFIIIC102general transcription factor IIIC|TFIIICgamma|TFIIIC 102 kDa subunit|102 kD
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:GTF3C3 (NP_036218, 112 a.a. ~ 215 a.a) partial recombinant protein with GST tag.TPEQPTAGDVFVLEMVLNRETKKMMKEKRPRSKLPRALRGLMGEANIRFARGEREEAILMCMEIIRQAPLAYEPFSTLAMIYEDQGDMEKSLQFELIAAHLNP
- Tested Applications:RNA Inhibition
- Purification:IgG purified
- Cat. No.:103336-656
- Supplier no.:H00009330-M02
Specifications
About this item
The gtf3c3 Antibody (3D9) from Novus Biologicals is a mouse monoclonal antibody to gtf3c3. This antibody reacts with human. The gtf3c3 Antibody (3D9) has been validated for the following applications: Western Blot, ELISA, RNA Inhibition.
Type: Primary
Antigen: gtf3c3
Clonality: Monoclonal
Clone: 3D9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human