To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:GSTA2
- Clonality:Monoclonal
- Clone:3D4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:2939
- Antigen Synonyms:GST HA subunit 2|class alpha|glutathione S-aryltransferase A2|glutathione S-transferase 2|MGC10525|liver GST2|GST|GTH2|S-(hydroxyalkyl)glutathione lyase A2|GST2glutathione S-alkyltransferase A2|glutathione S-transferase A2|GST class-alpha member 2|glutathione S-aralkyltransferase A2|EC 2.5.1.18|GSTA2-2|HA subunit 2|2|GTA2|glutathione S-transferase alpha 2|GST-gamma
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:GSTA2 (AAH02895, 1 a.a. - 100 a.a.) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MAEKPKLHYSNIRGRMESIRWLLAAAGVEFEEKFIKSAEDLDKLRNDGYLMFQQVPMVEIDGMKLVQTRAILNYIASKYNLYGKDIKEKALIDMYIEGIA
- Purification:IgG purified
- Cat. No.:103327-608
- Supplier no.:H00002939-M05
Specifications
About this item
The GSTA2 Antibody (3D4) from Novus Biologicals is a mouse monoclonal antibody to GSTA2. This antibody reacts with human. The GSTA2 Antibody (3D4) has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence.
Type: Primary
Antigen: GSTA2
Clonality: Monoclonal
Clone: 3D4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human