To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:GPR84
- Clonality:Monoclonal
- Clone:5B7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:53831
- Antigen Synonyms:GPCR4|G protein-coupled receptor 84|EX33Inflammation-related G-protein coupled receptor EX33|inflammation-related G protein-coupled receptor EX33|G-protein coupled receptor 84
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:GPR84 (NP_065103, 208 a.a. ~ 317 a.a) partial recombinant protein with GST tag.AAQALDQYKLRQASIHSNHVARTDEAMPGRFQELDSRLASGGPSEGISSEPVSAATTQTLEGDSSEVGDQINSKRAKQMAEKSPPEASAKAQPIKGARRAPDSSSEFGK
- Purification:Ascites
- Cat. No.:103344-598
- Supplier no.:H00053831-M02
Specifications
About this item
The GPR84 Antibody (5B7) from Novus Biologicals is a mouse monoclonal antibody to GPR84. This antibody reacts with human. The GPR84 Antibody (5B7) has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: GPR84
Clonality: Monoclonal
Clone: 5B7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human