To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:GPIP137
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:4076
- Antigen Synonyms:cell cycle associated protein 1|membrane component|surface marker 1|GPI-p137|Cytoplasmic activation- and proliferation-associated protein 1|activation/proliferation-associated protein 1|chromosome 11|RNG105GPIAP1|Cell cycle-associated protein 1|Membrane component chromosome 11 surface marker 1|GPI-anchored membrane protein 1GPIP137|caprin 1|caprin-1|RNA granule protein 105|p137GPI|M11S1GPI-anchored protein p137|cytoplasmic activation/proliferation-associated protein-1
- Storage Buffer:0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Concentration:0.25 mg/ml
- Immunogen:Recombinant protein encompassing a sequence within the center region of human GPIP137. The exact sequence is proprietary.
- Purification:Immunogen affinity purified
- Cat. No.:102708-402
- Supplier no.:NBP2-16712
Specifications
About this item
The GPIP137 Antibody from Novus Biologicals is a rabbit polyclonal antibody to GPIP137. This antibody reacts with human. The GPIP137 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry.
Type: Primary
Antigen: GPIP137
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human