To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:GNIP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:81786
- Antigen Synonyms:RNF90tripartite motif protein TRIM7|tripartite motif containing 7|tripartite motif-containing 7|Glycogenin-interacting protein|GNIPRING finger protein 90|tripartite motif-containing protein 7
- Storage Buffer:PBS (pH 7.2) and 40% Glycerol
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Immunogen:This antibody was developed against Recombinant Protein corresponding to amino acids:LRVLKKELEDCEVFRSTEKKESKELLKQMAAEQEKVGAEFQALRAFLVEQEGRLLGRLEELSREVAQKQNENLAQLGVEITQLSKLSSQI
- Purification:Immunogen affinity purified
- Cat. No.:103277-222
- Supplier no.:NBP1-89751
Specifications
About this item
The GNIP Antibody from Novus Biologicals is a rabbit polyclonal antibody to GNIP. This antibody reacts with human. The GNIP Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: GNIP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human