To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:GNB3
- Clonality:Monoclonal
- Clone:M1-1-1D5
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2a kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:2784
- Antigen Synonyms:Transducin beta chain 3|beta polypeptide 3|G protein|guanine nucleotide binding protein (G protein)|guanine nucleotide-binding protein G(I)/G(S)/G(T) subunit beta-3|guanine nucleotide-binding protein G(I)/G(S)/G(T) beta subunit 3|beta-3 subunit|GTP-binding regulatory protein beta-3 chain|hypertension associated protein
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:GNB3 (AAH02454, 1 a.a. - 340 a.a.) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MGEMEQLRQEAEQLKKQIADARKACAGVTLAELVSGLEVVGRVQMRTRRTLRGHLAKIYAMHWATDSKLLVSASQDGKLIVWDSYTTNKVHAIPLRSSWVMTCAYAPSGNFVACGGLDNMCSIYNLKSREGNVKVSRELSAHTGYLSCCRFLDDNNIVTSSGDTTCALWDIETGQQKTVFVGHTGDCMSLAVSPDFNLFISGACDASAKLWDVREGTCRQTFTGHESDINAICFFPNGEAICTGSDDASCRLFDLRADQELICFSHESIICSITSVAFSLSGRLLFAGYDDFNCNVWDSMKSERVGILSGHDNRVSCLGVTADGMAVATGSWDSFLKIWN
- Purification:IgG purified
- Cat. No.:103327-346
- Supplier no.:H00002784-M01
Specifications
About this item
The GNB3 Antibody (M1-1-1D5) from Novus Biologicals is a mouse monoclonal antibody to GNB3. This antibody reacts with human. The GNB3 Antibody (M1-1-1D5) has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence.
Type: Primary
Antigen: GNB3
Clonality: Monoclonal
Clone: M1-1-1D5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human