To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Glutamine Synthetase
- Clonality:Monoclonal
- Clone:3B6
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:2752
- Antigen Synonyms:PIG43|Glutamate--ammonia ligase|glutamine synthetase|proliferation-inducing protein 43|cell proliferation-inducing protein 59|GLNSPIG59|Glutamate decarboxylase|glutamate-ammonia ligase|GS|EC 4.1.1.15|glutamate-ammonia ligase (glutamine synthase)|EC 6.3.1.2|glutamine synthase
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:GLUL (NP_002056 274 a.a. - 373 a.a.) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. IEKLSKRHQYHIRAYDPKGGLDNARRLTGFHETSNINDFSAGVANRSASIRIPRTVGQEKKGYFEDRRPSANCDPFSVTEALIRTCLLNETGDEPFQYKN
- Purification:IgG purified
- Cat. No.:103327-296
- Supplier no.:H00002752-M02
The Glutamine Synthetase Antibody (3B6) from Novus Biologicals is a mouse monoclonal antibody to Glutamine Synthetase. This antibody reacts with human, mouse, rat. The Glutamine Synthetase Antibody (3B6) has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: Glutamine Synthetase
Clonality: Monoclonal
Clone: 3B6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse, Rat