To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:gamma-2 Tubulin
- Clonality:Monoclonal
- Clone:4F6
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2a kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:27175
- Antigen Synonyms:tubulin gamma-2 chain|MGC131994|tubulin|gamma-2-tubulin|gamma 2
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:TUBG2 (NP_057521, 1 a.a. ~ 101 a.a) partial recombinant protein with GST tag.MPREIITLQLGQCGNQIGFEFWKQLCAEHGISPEGIVEEFAT EGTDRKDVFFYQADDEHYIPRAVLLDLEPRVIHSILNSPYAK LYNPENIYLSEHGGGA
- Purification:IgG purified
- Cat. No.:103342-652
- Supplier no.:H00027175-M01
Specifications
About this item
The gamma-2 Tubulin Antibody (4F6) from Novus Biologicals is a mouse monoclonal antibody to gamma-2 Tubulin. This antibody reacts with human. The gamma-2 Tubulin Antibody (4F6) has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence.
Type: Primary
Antigen: gamma-2 Tubulin
Clonality: Monoclonal
Clone: 4F6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human