To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:gamma Sarcoglycan
- Clonality:Monoclonal
- Clone:3C5
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG2a kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:6445
- Antigen Synonyms:A4|sarcoglycan|gamma (35kD dystrophin-associated glycoprotein)|SCG3LGMD2C|MGC130048|gamma sarcoglycan|35 kDa dystrophin-associated glycoprotein|SCARMD2|DAGA4,35kD dystrophin-associated glycoprotein|MAM|DMDA1|gamma-sarcoglycan|gamma (35kDa dystrophin-associated glycoprotein)|DMDA|TYPE|Gamma-SG|35DAG
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:SGCG (NP_000222.1 191 a.a. - 290 a.a.) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. DLRLESPTRSLSMDAPRGVHIQAHAGKIEALSQMDILFHSSDGMLVLDAETVCLPKLVQGTWGPSGSSQSLYEICVCPDGKLYLSVAGVSTTCQEHSHIC
- Purification:IgG purified
- Cat. No.:103310-076
- Supplier no.:H00006445-M02
Specifications
About this item
The gamma Sarcoglycan Antibody (3C5) from Novus Biologicals is a mouse monoclonal antibody to gamma Sarcoglycan. This antibody reacts with human. The gamma Sarcoglycan Antibody (3C5) has been validated for the following applications: Western Blot, ELISA, Immunoprecipitation.
Type: Primary
Antigen: gamma Sarcoglycan
Clonality: Monoclonal
Clone: 3C5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human