To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:FIH-1/HIF-1AN
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 405
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:55662
- Antigen Synonyms:DKFZp762F1811|factor inhibiting HIF1|Factor inhibiting HIF-1|FIH1FIH-1|hypoxia inducible factor 1|EC 1.14.11.16|FLJ22027|peptide-aspartate beta-dioxygenase|alpha subunit inhibitor|FLJ20615|Hypoxia-inducible factor asparagine hydroxylase|hypoxia-inducible factor 1-alpha inhibitor
- Storage Buffer:50mM Sodium Borate
- Storage Temperature:Store at 4C in the dark.
- Immunogen:A synthetic peptide made to the C-terminal region of human Factor Inhibiting HIF-1 (between residues 300 and the C-terminus). [UniProt# Q9NWT6]
- Purification:Immunogen affinity purified
- Cat. No.:103358-014
Specifications
About this item
The FIH-1 / HIF-1AN Antibody [Alexa Fluor« 405] from Novus Biologicals is a rabbit polyclonal antibody to FIH-1 / HIF-1AN. This antibody reacts with human, mouse, rat. The FIH-1 / HIF-1AN Antibody [Alexa Fluor« 405] has been validated for the following applications: Western Blot, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Type: Primary
Antigen: FIH-1/HIF-1AN
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 405
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat