To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:DNA Polymerase epsilon subunit 3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:0.05 mg
- Environmentally Preferable:
- Gene ID:54107
- Antigen Synonyms:DNA polymerase epsilon p17 subunit|DNA polymerase II subunit 3|CHRAC-17|histone fold protein CHRAC17|CHARAC17arsenic transactivated protein|DNA polymerase epsilon subunit p17|epsilon 3 (p17 subunit)|HuCHRAC17|asTP|EC 2.7.7.7|polymerase (DNA directed)|CHRAC17Ybl1|Chromatin accessibility complex 17 kDa protein|Arsenic-transactivated protein|DNA polymerase epsilon subunit 3|p17|YBL1
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:POLE3 (AAH03166.1, 1 a.a. ~ 147 a.a) full-length human protein.MAERPEDLNLPNAVITRIIKEALPDGVNISKEARSAISRAASVFVLYATSCANNFAMKGKRKTLNASDVLSAMEEMEFQRFVTPLKEALEAYRREQKGKKEASEQKKKDKDKKTDSEEQDKSRDEDNDEDEERLEEEEQNEEEEVDN
- Purification:Protein A purified
- Cat. No.:103344-676
- Supplier no.:H00054107-B02P
Specifications
About this item
The DNA Polymerase epsilon subunit 3 Antibody from Novus Biologicals is a mouse polyclonal antibody to DNA Polymerase epsilon subunit 3. This antibody reacts with human. The DNA Polymerase epsilon subunit 3 Antibody has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence.
Type: Primary
Antigen: DNA Polymerase epsilon subunit 3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human