To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Cytochrome c
- Clonality:Monoclonal
- Clone:7H8.2C12
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2b kappa
- Reactivity:Drosophila,Pig,Rabbit,Horse,Mammals,Human,Dog,Rat,Mouse
- Western Blot:Yes
- Size:0.025 mg
- Environmentally Preferable:
- Gene ID:54205
- Antigen Synonyms:cytochrome c|CYCHCS|somatic|THC4
- Storage Buffer:PBS containing 0.5% BSA
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Concentration:0.5 mg/ml
- Immunogen:Synthetic peptides corresponding to amino acids 1-80, 81-104 and 66-104 of pigeon CYT were used as the immunogen (Jemmerson et al. 1991). The antibody recognizes an epitope within amino acids 93-104 of pigeon cytochrome C based on competitive ELISA results (Jemmerson et al. 1991).
- Purification:Protein G purified
- Cat. No.:103359-312
Specifications
About this item
The Cytochrome c Antibody (7H8.2C12) from Novus Biologicals is a mouse monoclonal antibody to Cytochrome c. This antibody reacts with human, mouse, rat, porcine, canine, drosophila, equine, mammal, rabbit. The Cytochrome c Antibody (7H8.2C12) has been validated for the following applications: Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: Cytochrome c
Clonality: Monoclonal
Clone: 7H8.2C12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human, Mouse, Rat, Porcine, Canine, Drosophila, Equine, Mammal, Rabbit