To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Collagen II
- Clonality:Monoclonal
- Clone:3H1-F9
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:1280
- Antigen Synonyms:chondrocalcin|collagen type 2|Collagen-2|alpha 1|cartilage collagen|collagen II|collagen|type II|Alpha-1 type II collagen|SEDC|AOM|Collagen 2|collagen alpha-1(II) chain|spondyloepiphysealdysplasia|alpha-1 polypeptide|alpha 1 (primary osteoarthritis|ANFH|COL11A3|MGC131516|congenital)
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:COL2A1 (AAH07252, 1 a.a. - 268 a.a.) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MSAFAGLGPREKGPDPLQYMRADQAAGGLRQHDAEVDATLKSLNNQIESIRSPEGSRKNPARTCRDLKLCHPEWKSGDYWIDPNQGCTLDAMKVFCNMETGETCVYPNPANVPKKNWWSSKSKEKKHIWFGETINGGFHFSYGDDNLAPNTANVQMTFLRLLSTEGSQNITYHCKNSIAYLDEAAGNLKKALLIQGSNDVEIRAEGNSRFTYTALKDGCTKHTGKWGKTVIEYRSQKTSRLPIIDIAPMDIGGPEQEFGVDIGPVCFL
- Purification:IgG purified
- Cat. No.:103324-162
- Supplier no.:H00001280-M01
Specifications
About this item
The Collagen II Antibody (3H1-F9) from Novus Biologicals is a mouse monoclonal antibody to Collagen II. This antibody reacts with human. The Collagen II Antibody (3H1-F9) has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: Collagen II
Clonality: Monoclonal
Clone: 3H1-F9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human