To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:CKII alpha prime polypeptide
- Clonality:Monoclonal
- Clone:1E8.
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2a kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:1459
- Antigen Synonyms:CK2A2|casein kinase 2|casein kinase II subunit alpha'|EC 2.7.11.1|EC 2.7.11|FLJ43934|CK II alpha'|CSNK2A1|alpha prime polypeptide
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:CSNK2A2 (NP_001887.1, 1 a.a. - 100 a.a.) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MPGPAAGSRARVYAEVNSLRSREYWDYEAHVPSWGNQDDYQLVRKLGRGKYSEVFEAINITNNERVVVKILKPVKKKKIKREVKILENLRGGTNIIKLID
- Purification:IgG purified
- Cat. No.:103324-526
- Supplier no.:H00001459-M03
Specifications
About this item
The CKII alpha prime polypeptide Antibody (1E8.) from Novus Biologicals is a mouse monoclonal antibody to CKII alpha prime polypeptide. This antibody reacts with human. The CKII alpha prime polypeptide Antibody (1E8.) has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence.
Type: Primary
Antigen: CKII alpha prime polypeptide
Clonality: Monoclonal
Clone: 1E8.
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human