To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Choline Acetyltransferase/ChAT
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:1103
- Antigen Synonyms:EC 2.3.1|CMS1A2|EC 2.3.1.6|choline O-acetyltransferase|acetyl CoA:choline O-acetyltransferase|Choline acetylase|CHOACTase|ChAT|choline acetyltransferase|CMS1A
- Storage Buffer:0.5 mg/ml Tris (pH 7.3) and 0.5% BSA
- Storage Temperature:Store at -20C. Avoid freeze-thaw cycles.
- Concentration:0.5 mg/ml
- Immunogen:Peptide with sequence CKEKATRPSQGHQP corresponding to C-Terminus according to NP_066264.3, NP_065574.3, NP_001136405.1.
- Purification:Immunogen affinity purified
- Cat. No.:103295-184
- Supplier no.:NB300-910
Specifications
About this item
The Choline Acetyltransferase / ChAT Antibody from Novus Biologicals is a goat polyclonal antibody to Choline Acetyltransferase / ChAT. This antibody reacts with human. The Choline Acetyltransferase / ChAT Antibody has been validated for the following applications: Western Blot, Peptide ELISA.
Type: Primary
Antigen: Choline Acetyltransferase/ChAT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human