To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Symbol:GIPC1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:0.05 mg
- Environmentally Preferable:
- Gene ID:10755
- Antigen Synonyms:C19orf3chromosome 19 open reading frame 3|GIPCRGS-GAIP-interacting protein|MGC15889|RGS19IP1IIP-1|regulator of G-protein signalling 19 interacting protein 1|NIP|GIPC PDZ domain containing family|TIP-2RGS19-interacting protein 1|MGC3774|GLUT1 C-terminal binding protein|Tax interaction protein 2|GLUT1CBP|SEMCAP|GAIP C-terminus-interacting protein|PDZ domain-containing protein GIPC1|SYNECTIIN|member 1|IGF-1 receptor interacting protein 1|SYNECTIN
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:GIPC1 (NP_005707.1, 1 a.a. ~ 333 a.a) full-length human protein.MPLGLGRRKKAPPLVENEEAEPGRGGLGVGEPGPLGGGGSGGPQMGLPPPPPALRPRLVFHTQLAHGSPTGRIEGFTNVKELYGKIAEAFRLPTAEVMFCTLNTHKVDMDKLLGGQIGLEDFIFAHVKGQRKEVEVFKSEDALGLTITDNGAGYAFIKRIKEGSVIDHIHLISVGDMIEAINGQSLLGCRHYEVARLLKELPRGRTFTLKLTEPRKAFDMISQRSAGGRPGSGPQLGTGRGTLRLRSRGPATVED
- Purification:Protein A purified
- Cat. No.:103339-528
- Supplier no.:H00010755-B01P
The GIPC1 Antibody from Novus Biologicals is a mouse polyclonal antibody to GIPC1. This antibody reacts with human. The GIPC1 Antibody has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: GIPC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human