To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:GIMAP2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:0.05 mg
- Environmentally Preferable:
- Gene ID:26157
- Antigen Synonyms:Immunity-associated protein 2|IMAP family member 2|immunity associated protein 2|DKFZp586D0824|IMAP2GTPase IMAP family member 2|GTPase|HIMAP2|MGC24275
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:GIMAP2 (NP_056475.1, 1 a.a. ~ 337 a.a) full-length human protein.MDQNEHSHWGPHAKGQCASRSELRIILVGKTGTGKSAAGNSILRKQAFESKLGSQTLTKTCSKSQGSWGNREIVIIDTPDMFSWKDHCEALYKEVQRCYLLSAPGPHVLLLVTQLGRYTSQDQQAAQRVKEIFGEDAMGHTIVLFTHKEDLNGGSLMDYMHDSDNKALSKLVAACGGRICAFNNRAEGSNQDDQVKELMDCIEDLLMEKNGDHYTNGLYSLIQRSKCGPVGSDERVKEFKQSLIKYMETQRSYTA
- Purification:Protein A purified
- Cat. No.:103342-156
- Supplier no.:H00026157-B01P
Specifications
About this item
The GIMAP2 Antibody from Novus Biologicals is a mouse polyclonal antibody to GIMAP2. This antibody reacts with human. The GIMAP2 Antibody has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: GIMAP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human