To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Symbol:FoxC2
- Clonality:Monoclonal
- Clone:3H5
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b kappa
- Reactivity:Human,Rat
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:2303
- Antigen Synonyms:Forkhead-related protein FKHL14|mesenchyme forkhead 1|Transcription factor FKH-14|Drosophila|homolog-like 14|mesenchyme forkhead 1)|Mesenchyme fork head protein 1|forkhead|MFH-1|FKHL14LD|MFH-1 protein|forkhead box C2 (MFH-1|MFH1forkhead box protein C2
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:FOXC2 (NP_005242 156 a.a. - 256 a.a.) full length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. FENGSFLRRRRRFKKKDVSKEKEERAHLKEPPPAASKGAPATPHLADAPKEAEKKVVIKSEAASPALPVITKVETLSPESALQGSPRSAASTPAGSPDGSL
- Purification:IgG purified
- Cat. No.:103326-564
- Supplier no.:H00002303-M05
The FoxC2 Antibody (3H5) from Novus Biologicals is a mouse monoclonal antibody to FoxC2. This antibody reacts with human, rat. The FoxC2 Antibody (3H5) has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: FoxC2
Clonality: Monoclonal
Clone: 3H5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human, Rat