To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Symbol:Fgr
- Clonality:Monoclonal
- Clone:3G10
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2a kappa
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:2268
- Antigen Synonyms:Gardner-Rasheed feline sarcoma viral (v-fgr) oncogene homolog|c-src-2 proto-oncogene|EC 2.7.10|p55c-fgr|c-fgr|Proto-oncogene c-Fgr|p55-Fgr|FLJ43153|p55-c-fgr protein|tyrosine-protein kinase Fgr|proto-oncogene tyrosine-protein kinase FGR|p58c-fgr|MGC75096|c-src2|EC 2.7.10.2|SRC2c-fgr protooncogene
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:FGR (AAH64382, 1 a.a. - 90 a.a.) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MGCVFCKKLEPVATAKEDAGLEGDFRSYGAADHYGPDPTKARPASSFAHIPNYSNFSSQAINPGFLDSGTIRGVSGIGVTLFIALYDYEA
- Purification:IgG purified
- Cat. No.:103326-426
- Supplier no.:H00002268-M01
The Fgr Antibody (3G10) from Novus Biologicals is a mouse monoclonal antibody to Fgr. This antibody reacts with human, mouse, rat. The Fgr Antibody (3G10) has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: Fgr
Clonality: Monoclonal
Clone: 3G10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human, Mouse, Rat