To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Symbol:FDX1
- Clonality:Monoclonal
- Clone:1E7.
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:2230
- Antigen Synonyms:mitochondrial adrenodoxin|ferredoxin 1|FDX|mitochondrial|LOH11CR1D|ferredoxin-1|ADXhepatoredoxin|adrenodoxin|Hepatoredoxin|Adrenal ferredoxin
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:FDX1 (NP_004100.1 85 a.a. - 183 a.a.) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. VGDSLLDVVVENNLDIDGFGACEGTLACSTCHLIFEDHIYEKLDAITDEENDMLDLAYGLTDRSRLGCQICLTKSMDNMTVRVPETVADARQSIDVGKT
- Purification:IgG purified
- Cat. No.:103326-304
- Supplier no.:H00002230-M01
The FDX1 Antibody (1E7.) from Novus Biologicals is a mouse monoclonal antibody to FDX1. This antibody reacts with human. The FDX1 Antibody (1E7.) has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: FDX1
Clonality: Monoclonal
Clone: 1E7.
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human