To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:FBXL7
- Clonality:Monoclonal
- Clone:2G10
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:23194
- Antigen Synonyms:F-box and leucine-rich repeat protein 7F-box protein FBL6/FBL7|KIAA0840|F-box/LRR-repeat protein 7|FBL7FBL6F-box protein Fbl7
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:FBXL7 (NP_036436, 392 a.a. ~ 490 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.HGVEYLAKNCTKLKSLDIGKCPLVSDTGLECLALNCFNLKRLSLKSCESITGQGLQIVAANCFDLQTLNVQDCEVSVEALRFVKRHCKRCVIEHTNPA
- Purification:IgG purified
- Cat. No.:103341-078
- Supplier no.:H00023194-M01
Specifications
About this item
The FBXL7 Antibody (2G10) from Novus Biologicals is a mouse monoclonal antibody to FBXL7. This antibody reacts with human. The FBXL7 Antibody (2G10) has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: FBXL7
Clonality: Monoclonal
Clone: 2G10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human