To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:ETV5/ERM
- Clonality:Monoclonal
- Clone:7C10
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:2119
- Antigen Synonyms:ETS translocation variant 5|ets variant gene 5 (ets-related molecule)|ets-related molecule|ERMEts-related protein ERM|ets variant 5
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:ETV5 (NP_004445, 181 a.a. - 290 a.a.) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. LPEPGPQQQTFAVPRPPHQPLQMPKMMPENQYPSEQRFQRQLSEPCHPFPPQPGVPGDNRPSYHRQMSEPIVPAAPPPPQGFKQEYHDPLYEHGVPGMPGPPAHGFQSPM
- Purification:IgG purified
- Cat. No.:103325-986
- Supplier no.:H00002119-M02
Specifications
About this item
The ETV5 / ERM Antibody (7C10) from Novus Biologicals is a mouse monoclonal antibody to ETV5 / ERM. This antibody reacts with human, mouse, rat. The ETV5 / ERM Antibody (7C10) has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: ETV5/ERM
Clonality: Monoclonal
Clone: 7C10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse, Rat