To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:ENPP-2/Autotaxin
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 μl
- Environmentally Preferable:
- Gene ID:5168
- Antigen Synonyms:PDNP2NPP2|ATXFLJ26803|Extracellular lysophospholipase D|AUTOTAXIN|autotaxin-t|EC 3.1.4.39|ectonucleotide pyrophosphatase/phosphodiesterase 2|LysoPLD|E-NPP 2|ATX-X|ectonucleotide pyrophosphatase/phosphodiesterase family member 2|PD-IALPHA|plasma lysophospholipase D|phosphodiesterase I/nucleotide pyrophosphatase 2
- Storage Buffer:PBS (pH 7.0) and 20% Glycerol
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Recombinant protein encompassing a sequence within the center region of human Autotaxin. The exact sequence is proprietary.
- Purification:Immunogen affinity purified
- Cat. No.:102707-716
- Supplier no.:NBP2-16339
Specifications
About this item
The ENPP-2 / Autotaxin Antibody from Novus Biologicals is a rabbit polyclonal antibody to ENPP-2 / Autotaxin. This antibody reacts with human. The ENPP-2 / Autotaxin Antibody has been validated for the following applications: Immunocytochemistry / Immunofluorescence.
Type: Primary
Antigen: ENPP-2/Autotaxin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human