To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:ELF5
- Clonality:Monoclonal
- Clone:3D10
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:2001
- Antigen Synonyms:ETS-related transcription factor Elf-5|Epithelium-restricted ESE-1-related Ets factor|ESE2ESE-2|Epithelium-specific Ets transcription factor 2|E74-like factor 5|E74-like factor 5 (ets domain transcription factor)
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:ELF5 (NP_938195.1, 166 a.a. - 263 a.a.) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. SRTSLQSSHLWEFVRDLLLSPEENCGILEWEDREQGIFRVVKSEALAKMWGQRKKNDRMTYEKLSRALRYYYKTGILERVDRRLVYKFGKNAHGWQED
- Purification:IgG purified
- Cat. No.:103325-664
- Supplier no.:H00002001-M01
Specifications
About this item
The ELF5 Antibody (3D10) from Novus Biologicals is a mouse monoclonal antibody to ELF5. This antibody reacts with human. The ELF5 Antibody (3D10) has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: ELF5
Clonality: Monoclonal
Clone: 3D10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human