To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Symbol:EIF2 beta
- Clonality:Monoclonal
- Clone:2F3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:8894
- Antigen Synonyms:38kDa|EIF2|eukaryotic translation initiation factor 2|eukaryotic initiation factor 2-beta|subunit 2 beta|EIF2beta|EIF2B|Eukaryotic translation initiation factor 2 subunit beta|eukaryotic translation initiation factor 2 beta|subunit 2 (beta|38kD )|eIF-2-beta|MGC8508|eukaryotic translation initiation factor 2 subunit 2|DKFZp686L18198
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:EIF2S2 (NP_003899.2, 1 a.a. - 100 a.a.) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MSGDEMIFDPTMSKKKKKKKKPFMLDEEGDTQTEETQPSETKEVEPEPTEDKDLEADEEDTRKKDASDDLDDLNFFNQKKKKKKTKKIFDIDEAEEGVKD
- Purification:IgG purified
- Cat. No.:103335-918
- Supplier no.:H00008894-M09
The EIF2 beta Antibody (2F3) from Novus Biologicals is a mouse monoclonal antibody to EIF2 beta. This antibody reacts with human, mouse, rat. The EIF2 beta Antibody (2F3) has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: EIF2 beta
Clonality: Monoclonal
Clone: 2F3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse, Rat